For Orbitrap data, you can filter reporter ion quantification spectra that have too much variability in the intensities of the reporter ions by using signal-to-noise values instead of intensities.
For Orbitrap data, you can filter reporter ion quantification spectra that have too much variability in the intensities of the reporter ions by using signal-to-noise values instead of intensities.