The Reporter Ions Quantifier node and the Precursor Ions Quantifier node do not display quantification information for Protein Groups.
The Reporter Ions Quantifier node and the Precursor Ions Quantifier node do not display quantification information for Protein Groups.