After the Reporter Ions Quantifier node and the Precursor Ions Quantifier node classify peptide groups, they calculate protein abundances for the samples from the associated peptide group abundances. They calculate a protein abundance as the simple summation of its associated and used peptide group abundances.
The nodes create the Abundance Counts column on the Proteins page, which shows the number of peptide group abundances used for calculating abundances.