Use the biological replicate study factor for experiments that include both technical and biological replicates. You can also use the biological replicate study factor to create a nested design study. Biological replicate study factors help you specify how to calculate p-values. You can set up these replicates as nested designs or non-nested designs.