Display the job queue - Proteome Discoverer Software - Display the job queue - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 User Guide
- Product Tree
- Desktop Software > Proteome Discoverer Software
- Ardia Platform > Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Choose Administration > Show Job Queue, or click the Show Job Queue,
. - The job queue opens on the Administration page of the application.
- To expand and view the details of the search, click the expand icon (+) to the left of the search name.
- The job queue uses different colors to indicate the type of workflow job and its execution state:
- Lighter green: Indicates successfully completed consensus workflows.
- Darker green: Indicates successfully completed processing workflows.
- Red: Indicates failed jobs.
- Dark gray: Indicates canceled jobs.
- Gray: Indicates informational messages and workflow processing that is in progress.
- Yellow: Indicates warning messages.
- Red: Indicates error messages.
- The job queue also uses colors to differentiate the messages that it issues during processing:
- Red: Error messages
- Yellow: Warning messages
- Gray: All other messages