The following table describes the shortcut menu commands that appear when you right-click the Workflow Tree pane of the Workflow Editor.
Command | Description |
|---|---|
Cut (CTRL+X) | Copies the selected node to the Clipboard and deletes it from the current Workflow Editor view. |
Copy (CTRL+C) | Copies the selected node to the Clipboard. |
Paste (CTRL+V) | Inserts a copied node into the current Workflow Editor view. |
Auto Layout | Automatically aligns the nodes in the workflow. |
Auto Number | Automatically numbers the nodes in a logical order, for example, the order that the nodes will process the data. |