You can right-click anywhere in any of the graphs displayed by the commands in the View menu to display a shortcut menu. Not all of the shortcut menu commands apply to all graphs; some graphs have unique commands. The following table lists the common commands in the graphs’ shortcut menu.
NOTE
The following figure shows common options between graphs and might differ depending on which graph is displayed. You might see more options in the menu.
Command | Description |
|---|---|
Show Position Tooltips | Displays information about a data point as a tooltip when you point to the data point in the plot. |
Zoom Out | Zooms out to the previous zoom perspective. |
Undo All Zoom/Pan | Zooms out completely. |
Copy | Copies the graph so that you can paste it into any business application document. |
Copy Points | Copies the graph data (but not the graph) so that you can paste it into a text document. |
Save As | Opens the Save As dialog box so that you can save the graph in EMF, PNG, JPG, GIF, TIFF, or BMP files. |
Save Points As | Opens the Save As dialog box so that you can save the graph data (but not the graph) as text in a TXT file. |