You can right-click anywhere on the pages of the result file to display a shortcut menu with the commands described in the following table.
Command | Description |
|---|---|
Copy With Headers | Copies the selected rows and the header captions to the Clipboard so that you can paste them into a third-party file application file, such as Excel. |
Copy | Copies the selected rows to the Clipboard so that you can paste them into a third-party file application, such as Excel. |
Clear Selection | Clears all selected items on a page of the result file. |
Enable Column Fixing | Moves a column to the leftmost position in the results report and keeps it stable and visible in that position, even when you scroll to the right through the rest of the columns. |
Collapse All Column Headers | Reduces the size of the column headers to their original size. Available only if you previously expanded the column headers with the Expand All Column Headers command. |
Expand All Column Headers | Expands the size of all column headers on a page of the result file at once so that you can see the labels of the different groups in a distribution map. |
Check Selected | Places a check mark next to the selected protein groups, proteins, peptide groups, PSMs, or precursor ions on the appropriate page of the results report.
|
Check All | Places a check mark next to all the selected protein groups, proteins, peptide groups, PSMs, or precursor ions on the appropriate page of the results report.
|
Uncheck Selected | Removes a check mark next to selected proteins or peptides on the Proteins page. |
Uncheck All | Removes a check mark next to the selected protein groups, proteins, peptide groups, PSMs, or precursor ions on the appropriate page of the results report.
|
Remove All Checkmarks in All Tables | Removes all check marks from all tables in the open result file. |
Add Tag | Add a tag to the selected row. |
Remove Tag | Remove a tag from the selected row. |
Set Tags | Add tags to multiple row at the same time. Hold CTRL + click or SHIFT + click to select the rows. |
Remove All Tags in All Tables | Removes tags from all of the tables in the open result file. |