The Tools menu contains the commands described in the following table.
Command | Function | Shortcut | Toolbar icon |
|---|---|---|---|
Tools > Open in FreeStyle | Starts FreeStyle, an application for visualizing and qualitatively analyzing mass spectrometry data. For more information, see Viewing Result Files Using FreeStyle. |
|
|
Tools > FASTA Database Utilities | Opens the FASTA Database Utilities dialog box. For parameter information, see Managing FASTA indexes. |
|
|
Tools > Create Bug Report | Opens the Proteome Discoverer Bug Reporter dialog box so that you can enter a product change request. |
| |
Tools > Mascot Communication Tester | Opens the Mascot Communication Tester dialog box and performs a series of tests to assist you in locating a problem if you cannot access the Mascot server. |
| |
Tools > CHIMERYS Communication Tester | Opens the CHIMERYS Communication Tester and Job Monitoring Tool dialog box. |
| |
Tools > Options | Opens the Options dialog box. For parameter information, see Options dialog box parameters. |