The toolbar is directly below the menu bar.
Toolbar icon | Function | Menu equivalent | Keyboard Shortcut |
|---|---|---|---|
Opens the New Study and Analysis dialog box for creating a new study. | File > New Study/Analysis |
| |
Opens the Open Study dialog box for selecting an existing study. | File > Open Study |
| |
Opens a result report.
| File > Open Result |
| |
Saves all the changes that you made to a report or a study. | File > Save | CTRL+S | |
Saves all open studies in the application.
| File > Save All | CTRL+SHIFT+S | |
Opens the Proteome Discoverer Start Page. | View > Start Page |
| |
Displays the Layout Selector area.
| View > Layout Selector | CTRL+L | |
Displays the Custom Tags Editor.
| View > Custom Tags Editor | CTRL+SHIFT+C | |
Opens the Display Filter pane for selecting display filters to filter data in the result file. | View > Display Filter | CTRL+F
| |
Displays the Report Item Distribution chart. | View > Distribution Charts | CTRL+SHIFT+D | |
Displays maps of the chemical pathways identified by the Wiki Pathways database. | View > Pathway Map |
| |
Displays Enrichment Chart View
| View > Enrichment Chart |
| |
Displays the Protein Identification Details view when you select a protein row. Displays the Peptide Spectrum Match Identification Details view when you select a peptide row. | View > Protein Details or Peptide Spectrum Match Details | CTRL+D | |
Displays the Chromatogram Traces view. | View > Chromatogram Traces View | CTRL+SHIFT+T | |
Displays the Fragment Match Spectrum view. | View > Fragment Match Spectrum | CTRL+SHIFT+F | |
Displays the Precursor Isotope Pattern view. | View > Precursor Isotope Pattern | CTRL+SHIFT+P | |
Displays the Sequence Comparison chart. | View > Sequence Comparison |
| |
Displays the Quantification Ratio Distributions chart. | View > Quan Ratio Distributions | CTRL+SHIFT+R | |
Displays the Trend chart. | View > Trend Chart |
| |
Displays the Quantification Channel Values chart. | View > Values | CTRL+SHIFT+N | |
Displays the Quantification Spectrum chart. | View > Quantification Spectrum | CTRL+SHIFT+Q | |
Displays the Result Summaries view.
| View > Summaries | CTRL+F | |
Opens the Workflow Editor for loading, editing, and saving workflow templates. | View > Workflow Editor |
| |
Displays the job queue. | Administration > Show Job Queue | CTRL+J | |
Opens the FASTA Files view for adding, removing, and modifying FASTA files. | Administration > Maintain FASTA Files |
| |
Opens the Chemical Modifications view to activate or deactivate available chemical modifications that are used in the search process, create new modifications, or edit existing modifications. | Administration > Maintain Chemical Modifications |
| |
Opens the Quantification Methods view, which lists all of the available methods for both precursor ion and reporter ion quantification. | Administration > Maintain Quantification Methods |
|
For information about customizing the toolbar, see Customizing the toolbar and Customize toolbar icons, fonts, and tooltips display.
NOTE
If you have the ProSightPD application, you may see additional icons in the Proteome Discoverer toolbar.