The application uses custom parsing rule C if the FASTA title line only contains the accession#. A typical FASTA title line, which matches this parsing rule, looks like this:
>143B_HUMAN
FASTA ID:
Accession#:143B_HUMAN
Description:143B_HUMAN
The application uses custom parsing rule C if the FASTA title line only contains the accession#. A typical FASTA title line, which matches this parsing rule, looks like this:
>143B_HUMAN
FASTA ID:
Accession#:143B_HUMAN
Description:143B_HUMAN