The application uses custom parsing rule B if the accession# and the description are separated by a space. A typical FASTA title line, which matches this parsing rule, looks like this:
>HP0001 hypothetical protein {Helicobacter pylori 26695}
FASTA ID:
  • Accession#:HP0001
  • Description:hypothetical protein {Helicobacter pylori 26695}