The Result Exporter node exports all the pages of a result file to separate tab-separated text files.
The following table describes the parameters for the Result Exporter node.
Parameter | Definition |
|---|---|
File Name | Specifies the name of the file to export the results to. |
Export Format | The data format in which the result data should be stored.
|
R-Friendly Headers | Generates headers with the appropriate characters when you export data to the R Project for Statistical Computing open source environment.
|