Post-processing nodes apply their functionality to the workflow after the nodes of the processing and other consensus nodes. Post-processing nodes process the results. Examples of tasks that can be handled in post-processing are:
- Exporting results to another format, such as a TXT.
- Transferring the generated results to another location or server.
You can add post-processing nodes only to the Post-Processing Nodes pane. Similarly, you cannot add nodes without post-processing functionality to the Post-Processing Nodes pane.
For information on the Scripting Node, see Custom script integration.