The Peptide Isoform Grouper node applies an alternative grouping of Peptide Spectrum Matches (PSMs) that considers the position of the measured modifications as an additional grouping criterium.
The node creates an additional table named “Isoform Groups,” within each isoform group, all PSMs have the same modifications at the same positions.
The Peptide Isoform Grouper node has no parameters.