The Reporter Ions Quantifier node is required for reporter ion quantification. The node includes reporter-ion-specific settings such as isolation interference and minimum average reporter ion S/N. It controls the peptides to be used for quantification on the basis of their uniqueness. It also has parameters for normalization, scaling, and imputation. It is responsible for the peptide-to-protein quantification rollup.
The default reporter ion quantification workflows include this quantification node.
The following table describes the parameters for the Reporter Ions Quantifier node.
Parameter | Definition |
|---|---|
General Quantification Settings |
|
Peptides to Use | Specifies the peptides to use for protein quantification.
This parameter adds the # Razor Peptides column to the Proteins page of the result report when you use razor peptides for quantification. |
Consider Protein Groups for Peptide Uniqueness | Defines peptide uniqueness on the basis of protein groups rather than individual proteins.
For best results, use the default. When you use larger protein (FASTA) databases, peptides commonly appear in multiple homologous proteins. If you set this parameter to False, the results might not contain many quantification results. |
Use Shared Quan Results | If set to True, the shared quan results are used for quantification; otherwise, they are not. A quan result is a Reporter Quan Spectrum in reporter quantification, and it is a consensus feature in precursor quantification. Quan results are shared if they are associated with multiple peptides. |
Reject Quan Results with Missing Channels | Determines whether to set all quantification values from all channels to zero if one or more of the quantification channels have a detected intensity of zero.
|
Reporter Quantification |
|
Reporter Abundance Based On | Specifies whether to use S/N values or intensity values as quantification channel values instead of ion reporter peak values in reporter ion quantification results.
The workflow fails when the spectra do not contain S/N values, as can happen with ion trap spectra. |
Apply Quan Value Corrections | Determines whether to apply a correction (for example, for isotopic or labeling impurities) to the raw quantification values.
|
Co-Isolation Threshold | Specifies a threshold for the amount of interference allowed by coisolation. Interference is the amount of ion current within the isolation window not originating from the identified peptide. Peptides with percentages higher than this threshold should not be used for quantification. The default value of 100% means that no peptide is excluded. This parameter is only used for reporter ion quantification. For more information on coisolation, see Excluding PSMs with high levels of coisolation. Range: 0–100; default: 50 |
Average Reporter S/N Threshold | Specifies the minimum average reporter S/N threshold value that determines which PSMs the application excludes from quantification. The application excludes all PSMs that have an average S/N value smaller than this threshold value. This parameter adds the Average Reporter S/N column, which displays average reporter S/N values, to the PSMs and Quan Spectra pages. The application calculates these values as the sum of S/N values found, divided by the number of defined mass tags. For detailed information about the calculation, see Filtering quantification data with average reporter ion signal-to-noise values. For the PSMs that this parameter excludes from quantification, the Quan Info column displays the notation Excluded by Method, and the Quan Usage columns displays the notation Not Used. Range: 0–no maximum; default: 0 |
CHIMERYS Coefficient Threshold | Only spectra with a CHIMERYS Coefficient above this threshold are considered for the quantification. The CHIMERYS Coefficient represents the contribution of the PSM to the MS2 spectrum. |
SPS Mass Matches [%] Threshold | This parameter specifies the percentage of all selected SPS masses that have to match a peptide fragment for considering this PSM in the quantification. That is, when there are ten SPS masses, then at least seven have to match a peptide fragment, otherwise the PSM is discarded from quantification. Default: 65 |
Minimum Channel Occupancy [%] Threshold | Only peptides with at least this percentage of non-zero channel values are considered for quantification. Range: 0 – 100; default: 0 |
Normalization and Scaling |
|
Normalization Mode | Specifies how to perform normalization to correct for experimental bias.
|
Proteins for Normalization | Specifies the name of the FASTA database containing the proteins that the normalization should be based on. The database can contain a single protein sequence or multiple protein sequences. This parameter is relevant only when you set the Normalization Mode parameter to Specific Protein Amount. |
Scaling Mode | Determines how Precursor Ions Quantifier node scales abundances. After it aggregates all the abundance values or optionally normalized abundance values per quantification channel, the node scales the abundance values of each channel as follows:
|
Exclude Peptides from Protein Quantification | |
For Normalization | Specifies how the normalization based on the summed abundance of all peptides and the RT-dependent normalization are done:
|
For Protein Roll-Up | Specifies how the roll-up of peptide abundances to protein level is done:
|
For Pairwise Ratios | Specifies which peptides to use to calculate protein ratios based on the pairwise peptide ratios mode:
|
Considered Peptide Modification | Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section. |
N-Terminal Considered Peptide Modification | Does not use peptides with the specified modification to calculate the normalization factors. Only effective when you set the Normalization Mode parameter to Total Peptide Amount. |
C-Terminal Considered Peptide Modification | Does not use peptides with the specified modification to calculate the normalization factors. Only effective when you set the Normalization Mode parameter to Total Peptide Amount. |
N-Terminal Considered Protein Modification | Does not use peptides with the specified modification to calculate the normalization factors. Only effective when you set the Normalization Mode parameter to Total Peptide Amount. |
C-Terminal Considered Protein Modification | Does not use peptides with the specified modification to calculate the normalization factors. Only effective when you set the Normalization Mode parameter to Total Peptide Amount. |
Quan Rollup and Hypothesis Testing |
|
Protein Ratio Calculation | Specifies how protein ratios are calculated.
|
Maximum Allowed Fold Change | Specifies the maximum fold change that can be reasonably expected for the observed ratios. Reports the quantification ratios based on the maximum values. Values greater than the value selected by this option are replaced by the maximum or minimum value. Range: 1–100 000; default: 100 With the default setting, ratios above 100 are set to 100, and ratios below 0.01 are set to 0.01. |
Imputation Mode | Specifies how missing values should be treated.
For more information on the Imputation Mode parameter, see Impute missing values. |
Hypothesis Test | Specifies how p-values are calculated for the reported quan ratios.
|
Quan Ratio Distributions |
|
1st Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the first highlight color. A ratio of 1.0 indicates no change. Range: 1.0–1000.0; default: 2 |
2nd Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the second highlight color. A ratio of 1.0 means no change. Range: 0.0–1000.0; default: 4 |
3rd Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the third highlight color. A ratio of 1.0 means no change. Range: 0.0–1000.0; default: 6 |
4th Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the fourth highlight color. Range: 0.0–1000.0; default: 8 |
5th Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the fifth highlight color. Range: 0.0–1000.0; default: 10 |
The following figure shows the five colors associated with the nth Fold Change Threshold parameters.