The Precursor Ions Quantifier node is required for precursor ion and label-free quantification. It controls the peptides to be used for quantification on the basis of their uniqueness. It can also set parameters for normalization, scaling, and imputation. It is responsible for the peptide-to-protein quantification rollup.
The default workflows for label-free quantification and precursor ion quantification include this quantification node.
The following table describes the parameters for the Precursor Ions Quantifier node.
Parameter | Definition |
|---|---|
General Quantification Settings |
|
Peptides to Use | Specifies the peptides to use for protein quantification.
This parameter adds the # Razor Peptides column to the Proteins page of the result report when you use razor peptides for quantification. |
Consider Protein Groups for Peptide Uniqueness | Defines peptide uniqueness on the basis of protein groups rather than individual proteins.
For best results, use the default parameter. When you use larger protein (FASTA) databases, peptides commonly appear in multiple homologous proteins. If you set this parameter to False, the results might not contain many quantification results. |
Use Shared Quan Results | If set to True, the shared quan results are used for quantification; otherwise, they are not. A quan result is a reporter quan spectrum in reporter quantification, and it is a consensus feature in precursor quantification. Quan results are shared if they are associated with multiple peptides. |
Reject Quan Results with Missing Channels | Determines whether to set all quantification values from all channels to zero if one or more of the quantification channels have a detected intensity of zero.
|
Precursor Abundance Based On | Specifies whether to use area or intensity to calculate precursor abundance in the quantification results.
|
Min. # Replicate Features [%] | Specifies the minimum percentage of replicate files that a feature must be detected in to be used. The application uses only the quantification values detected in a specified percentage of replicates. It determines which files belong to one replicate and whether there are quantification values for enough replicate files. If it does not find enough files, it excludes LC/MS features from the replicate group from quantification. This parameter affects only label-free quantification. Range: 0–100; default: 0 |
Normalization Mode | Specifies how to perform normalization to correct for experimental bias.
For more information on the Normalization Mode parameter, see Normalizing peptide groups and protein abundances. |
Proteins for Normalization | Specifies the name of the FASTA database containing the proteins that the normalization should be based on. The database can contain a single protein sequence or multiple protein sequences. This parameter is relevant only when you set the Normalization Mode parameter to Specific Protein Amount. |
Scaling Mode | Determines how Precursor Ions Quantifier node scales abundances. After it aggregates all the abundance values or optionally normalized abundance values per quantification channel, the node scales the abundance values of each channel as follows:
|
For Normalization | Specifies which peptides to use for deriving the normalization.
|
For Protein Roll-up | Specifies which peptides to use for calculation protein abundances.
|
For Pairwise Ratios | Specifies which peptides to use for calculation protein ratios based on the ‘Pairwise Ratio Based’ mode. The setting does not affect protein ratios directly calculated from protein abundances in the ‘Protein Abundance Based’ mode.
|
1. Considered Peptide Modification | Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section. |
2. Considered Peptide Modification | Does not use peptides with the specified modification to calculate the normalization factors. Only effective when you set the Normalization Mode parameter to Total Peptide Amount. |
3. Considered Peptide Modification | Does not use peptides with the specified modification to calculate the normalization factors. Only effective when you set the Normalization Mode parameter to Total Peptide Amount. |
N-Terminal Considered Protein Modification | Does not use peptides with the specified modification to calculate the normalization factors. Only effective when you set the Normalization Mode parameter to Total Peptide Amount. |
Protein Abundance Calculation | Specifies how protein abundances are calculated.
|
N for Top N | Specifies the number of distinct peptides used for calculating the protein abundance values from the connected peptides according to the Top N approach. The value is only relevant if ‘Top N Average’ is selected for calculating protein abundances. Minimum = 1; Maximum = (unchecked) |
Protein Ratio Calculation | Specifies how protein ratios are calculated.
|
Maximum Allowed Fold Change | Specifies the maximum fold change that can be reasonably expected for the observed ratios. Reports the quantification ratios based on the maximum values. Values greater than the value selected by this option are replaced by the maximum or minimum value. Range: 1–100 000; default: 100 With the default setting, ratios above 100 are set to 100, and ratios below 0.01 are set to 0.01. |
Imputation Mode | Specifies how to treat missing values.
For more information, see Impute missing values. |
Hypothesis Test | Specifies how p-values are calculated for the reported quan ratios.
|
1st Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the first highlight color. A ratio of 1.0 indicates no change. Range: 1.0–1000.0; default: 2 |
2nd Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the second highlight color. A ratio of 1.0 means no change. Range: 0.0–1000.0; default: 4 |
3rd Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the third highlight color. A ratio of 1.0 means no change. Range: 0.0–1000.0; default: 6 |
4th Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the fourth highlight color. Range: 0.0–1000.0; default: 8 |
5th Fold Change Threshold | Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the fifth highlight color. Range: 0.0–1000.0; default: 10 |
The following figure shows the five colors associated with the nth Fold Change Threshold parameters.