The application uses the amino acid symbols and mass values listed in the following tables.
Amino acid | One-letter | Three-letter | Monoisotopic | Average | Sum |
|---|---|---|---|---|---|
Glycine | G | Gly | 57.02146 | 57.05177 | C2H3NO |
Alanine | A | Ala | 71.03711 | 71.07855 | C3H5NO |
Serine | S | Ser | 87.03203 | 87.07796 | C3H5NO2 |
Proline | P | Pro | 97.05276 | 97.11623 | C5H7NO |
Valine | V | Val | 99.06841 | 99.13211 | C5H9NO |
Threonine | T | Thr | 101.04768 | 101.10474 | C4H7NO2 |
Cysteine | C | Cys | 103.00918 | 103.14464 | C3H5NOS |
Isoleucine | I | Ile | 113.08406 | 113.15890 | C6H11NO |
Generic Leu/lle | J | Jli | 113.08406 | 113.15890 | C6H11NO |
Leucine | L | Leu | 113.08406 | 113.15890 | C6H11NO |
Asparagine | N | Asn | 114.04293 | 114.10354 | C4H6N2O2 |
Aspartic Acid | D | Asp | 115.02694 | 115.08826 | C4H5NO3 |
Glutamine | Q | Gln | 128.05858 | 128.13032 | C5H8N2O2 |
Lysine | K | Lys | 128.09496 | 128.17358 | C6H12N2O |
Glutamic Acid | E | Glu | 129.04259 | 129.11504 | C5H7NO3 |
Methionine | M | Met | 131.04048 | 131.19820 | C5H9NOS |
Histidine | H | His | 137.05891 | 137.14062 | C6H7N3O |
Phenylalanine | F | Phe | 147.06841 | 147.17571 | C9H9NO |
Arginine | R | Arg | 156.10111 | 156.18707 | C6H12N4O |
Tyrosine | Y | Tyr | 163.06333 | 163.17512 | C9H9NO2 |
Tryptophan | W | Trp | 186.07931 | 186.21220 | C11H10N2O |
Amino acid | One-letter | Three-letter | Monoisotopic | Average mass | Sum |
|---|---|---|---|---|---|
Avrg. N/D | B | Bnd | 114.53494 | 114.59590 |
|
Avrg. Q/E | Z | Zqe | 128.55059 | 128.62326 |
|
Unknown acid (X) | X | Xxx | 0 | 0 | N/A |
Pyrrolysine | O | Pyl | 237.14773 | 237.29874 | C12H19N3O2 |
Seleno Cysteine | U | Sec | 150.95364 | 150.0369 | C3H5NOSe |