The post processing workflows do not change the data, but they change how it is displayed. The nodes used in these workflows are:
- Display Settings node—for information on using this node in a workflow, see Create and apply layouts in the Proteome Discoverer Help.
- Data Distributions node—for information on using this node in a workflow, see Generate data distribution maps in the Proteome Discoverer Help.
- Result Statistics—for information on using this node, which has no parameters, see The Result Statistics page in the Proteome Discoverer Help.
- Result Exporter—for information on using this node in a workflow, see the Result Exporter node in the Proteome Discoverer Help.
For information about the Scripting Node, which appears in the Post Processing Section, see Custom script integration.