In the consensus workflow, you can use these parameters of the MSF Files node to filter PSMs:
  • Maximum Delta Cn
  • Maximum Rank
  • Maximum Delta Mass
  • Score/Threshold parameters of the individual search node
Special Processing Workflow Nodes and Node Settings
No special nodes or node settings required.
Special Consensus Workflow Nodes and Node Settings
All consensus workflow templates include the MSF Files node.
Further Information
For information about these parameters, see the MSF Files node in the Proteome Discoverer Help.