You can also filter PSMs by using the Peptide and Protein Filter node in the consensus workflow. This node filters by score for the type of score specified. For information on this node, see the Peptide and Protein Filter nodein the Proteome Discoverer Help.