You can view result files in the FreeStyle application if it is installed on your system. This application is used to visualize and qualitatively analyze mass spectrometry data.
The Proteome Discoverer application includes a menu item to open result files the FreeStyle application. For information about using the FreeStyle application, refer to the Thermo FreeStyle User Guide.
NOTE
The result file and the raw data file must reside in the same directory.
Procedure
- Go to the Input Files tab.
- Select the file.
- From the Tools menu, select Open in FreeStyle.