The shortcut menu in the Mass Recalibration view provides options to copy or export the plot as an image or the underlying data.
The following table describes the commands in the shortcut menu of the Mass Recalibration view.
Command or Option | Description |
|---|---|
Show Position Tooltips | Displays the coordinates of the location of the mass error and the retention time of the landmark peptide. |
Zoom Out | Reduces the view. |
Undo All Zoom/Pan | Returns the Mass Recalibration view to the default view. |
Copy | Copies the data in the Mass Recalibration view. |
Image | Copies the data in the Mass Recalibration view as an image. |
Points | Copies the data in the Mass Recalibration view as text. |
Export | Exports the data in the Mass Recalibration view to a file. |
Image As | Exports the data in the Mass Recalibration view as an image in an Enhanced Metafile (EMF) file. |
Points As | Exports the data in the Mass Recalibration view as text in a TXT file. |