The following table describes the parameters on the Configuration page.
Parameter | Description |
|---|---|
Consensus WF page (Consensus Workflow Configuration) | Displays information about any configuration settings in the consensus workflow. |
Result Name | Displays the name of the .pdResult file. |
Result File | Displays the path and name of the .pdResult file. |
Description | Displays a brief description of the search. This information comes from the Description field of the Workflow Editor. |
Workflow Based on Template | Displays the name of the consensus workflow template used, if any. |
Creation Date | Displays the date that the search was performed. |
Created with Discoverer Version | Displays the version of the application that generated the results file. |
Configuration for: node_name | Lists the server configuration settings for any applicable nodes used in the consensus workflow. |
Processing WF page (Processing Workflow Configuration) | Displays information about any configuration settings in the processing workflow. |
Workflow Created with Discoverer Version | Displays the version of the application that generated the results file. |
Search Name | Displays the name of the search. |
Search Template | Displays the name of the processing workflow template used, if any. |
Search Description | Displays a brief description of the search that comes from the Description field of the Workflow Editor. |
Search Date | Displays the date that the search was performed. |
Configuration for: node_name | Lists the server configuration settings for any applicable nodes used in the processing workflow. |