The following table describes the parameters on the Quantification page.
Parameter | Description |
|---|---|
Consensus Step page (Consensus Step Quantification) | Displays information about the quantification consensus workflow. |
Result Name | Displays the name of the .pdResult file. |
Result File | Displays the path and name of the .pdResult file. |
Description | Displays a brief description of the search. This information comes from the Description field of the Workflow Editor. |
Workflow Based on Template | Displays the name of the consensus workflow template used, if any. |
Creation Date | Displays the date that the search was performed. |
Created with Discoverer Version | Displays the version of the application that generated the results file. |
Processing Node number | Displays the parameter settings of the Precursor Ions Quantifier or the Reporter Ions Quantifier node in the consensus workflow. |
Processing Step page (Processing Step Quantification) | Displays information about the quantification processing workflow. |
Result Name | Displays the name of the .pdResult file. |
Result File | Displays the path and name of the .pdResult file. |
Description | Displays a brief description of the search. This information comes from the Description field of the Workflow Editor. |
Workflow Based on Template | Displays the name of the consensus workflow template used, if any. |
Creation Date | Displays the date that the search was performed. |
Created with Discoverer Version | Displays the version of the application that generated the results file. |
Quantification Method | Displays the Quantification Method. |
Required Peptide Modification | Displays the modifications set for the search engine used. |
Quan Channels | Displays the quantification channels. |
Purity Correction Factors in [%] | Displays any purity correction factors used in the quantification. |
Processing Node | Displays the settings of the quantification nodes. |
Derived Values page (Derived Quantification Values) | Displays correction factors applied to the raw quantification values. |
Minimum Quan Values | Displays the settings of the Apply Quan Value Corrections parameter of the Reporter Ions Quantifier node. |
Correction for Experimental Bias | Displays the settings of the Normalization Mode parameter of the Precursor Ions Quantifier node or the Reporter Ions Quantifier node. |
Reported Ratios page (Ratio and Ratio Groups) | Displays the quantification ratio groups generated and the ratios grouped together, along with their associated files. |