Use the Quantification page to view the settings that you chose for these nodes:
- Minora Feature Detector node
- Reporter Ions Quantifier node and Precursor Ions Quantifier node
You can also use the page to view the settings in the Quantification Method Editor for both precursor ion or reporter ion quantification.
You must conduct a search with a workflow that includes a quantification node for this page to appear in the results report.
Procedure
- In the Result Summaries pane, select the Quantification tab.
Quantification page