The following table describes the parameters on the Validation page.
Parameter | Description |
|---|---|
Consensus Step page (Consensus Step Validation) | Displays the settings of the validation nodes used in the consensus workflow. |
Result Name | Displays the name of the .pdResult file. |
Result File | Displays the path and name of the .pdResult file. |
Description | Displays the names of the nodes in the consensus workflow. |
Workflow Based on Template | Displays the name of the template file used to generate the .pdResult file. |
Creation Date | Displays the date and time that the .pdResult file was generated. |
Workflow Created with Discoverer Version | Displays the version of the application that created the .pdResult file. |
Peptide Validator Nodes | Displays a list of the peptide validation nodes used in the workflow and their parameter settings. |
Processing Step page (Processing Step Validation) | Displays the settings of the validation nodes used in the processing workflow. |
Result Name | Displays the name of the .pdResult file. |
Result File | Displays the path and name of the MSF file. |
Description | Displays the names of the raw data file and the FASTA file used to generate the MSF file. |
Workflow Based on Template | Displays the name of the template file used to generate the MSF file. |
Creation Date | Displays the date and time that the MSF file was generated. |
Workflow Created with Discoverer Version | Displays the version of the application that created the MSF file. |
Filters page (Filters and Counts) | Displays the settings of the display filters used to filter the data in the .pdResult file, if any. It also displays the number of result items not filtered out by the Peptide in Protein Annotation node and the Protein Grouping node on each page of the .pdResult file. |
FDR Values page (FDR Values for Entire Result) | Displays the FDR values found for protein groups, proteins, peptide groups, and PSMs before the application applied any display filters. |