Display the Validation page - Display the Validation page - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Display the Validation page - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- In the Result Summaries pane, select the Validation tab.
- The Validation page opens in the Result Summaries pane. This example displays FDR values calculated with the concatenation calculation method.
- Validation Summary page