The following table describes the parameters on the Analysis Settings page.
Parameter | Description |
|---|---|
Consensus Step page (Consensus Step Workflow) | Summarizes the consensus workflow. |
Result Name | Displays the name of the .pdResult file. |
Result File | Displays the path and name of the .pdResult file. |
Description | Displays a brief description of the search. This information comes from the Description in the Workflow Editor. |
Workflow Based on Template | Displays the name of the consensus workflow template used, if any. |
Creation Date | Displays the date that the search was performed. |
Created with Discoverer Version | Displays the version of the application that generated the results file. |
Pipeline Tree | Displays the nodes used in the workflow in order. |
List of nodes | Displays the parameter settings for each node used in the consensus workflow. |
Workflow Messages | Displays a list of all workflow messages that the application generated when it executed the consensus workflow. |
Processing Step Letter page (Processing Step Workflow) | Summarizes each processing workflow. |
Result Name | Displays the name of the .pdResult file. |
Result File | Displays the path and name of the .pdResult file. |
Description | Displays a brief description of the search that comes from the Description field of the Workflow Editor. |
Workflow Based on Template | Displays the name of the processing workflow template used, if any. |
Creation Date | Displays the date that the search was performed. |
Created with Discoverer Version | Displays the version of the application that generated the results file. |
Pipeline Tree | Displays the nodes used in the workflow in order. |
List of nodes | Displays the parameter settings for each node used in the processing workflow. |
Workflow Messages | Displays a list of all workflow messages that the application generated when it executed the processing workflow. |