The Analysis Settings page of the Result Summaries pane provides a text version of the parameters that you set to perform a search. When you open a result file, you can use the Analysis Settings page to obtain information about the workflow that produced it.
The Analysis Settings page is divided into several categories containing information about the following:
- The name of the report file and the names of the MSF files used to generate the report file
- The version of the application used to generate the report file
- A summary of the currently active display filters, if any
- A summary of the number of result items in the Protein Groups, Proteins, Peptide Groups, PSMs, and MS/MS Spectrum Info categories in the report after applying the display filters
- A summary of the workflow used for generating the report file and the parameter settings
- A list of all workflow messages generated during report file generation
- A summary of the workflows used for generating each of the MSF files and the parameter settings
- A list of all workflow messages generated during MSF file generation
You can access the Analysis Settings page from an open results report. It appears in a dockable window.
Procedure
- In the Result Summaries pane, select the Analysis Settings tab.
Analysis Settings page