The following table lists the parameters in the Precursor Isotope Pattern view.
Parameter | Description |
|---|---|
Header | Displays the following:
|
Intensity [counts] (y axis) | Displays the scale of intensity of the precursor scan. |
m/z (x axis) | Displays the mass-to-charge ratio (m/z) of the precursor scan. |