The following table lists the parameters in the Fragment Match Spectrum view.
Parameter | Description |
|---|---|
Header | Displays the following:
|
Red vertical lines | a-, b-, and c-series ions matched in the spectrum. |
Blue vertical lines | x-, y-, and z-series ions matched in the spectrum. |
Intensity (counts) (y axis) | Displays the scale of the intensity of the product ions. |
m/z (x axis) | Displays the mass-to-charge ratio of the product ions, in daltons. |