The Chromatogram Traces view displays the following:
- The chromatographic profiles of individual features or peaks that the application uses to calculate label-free and precursor ion quantification results
- The exact peak and feature traces that are used to derive quantification values
In addition, you can separately display the single peaks that are subsumed in feature traces to provide an even more detailed view. Closely overlapping chromatograms (or traces) in the view indicate whether peptides eluted at the same time from the column and therefore are the same peptide. They confirm the validity of the quantification results and the origin of the consensus features from the same peptide. Conversely, peaks appearing far apart in the view indicate potential false positives.
NOTE
The Chromatogram Traces view is available only when you perform label-free quantification or precursor ion quantification, but what it displays in the view depends on the result file page that is open.
Alignment is a shift in the column during elution. It takes the pattern across different runs and shifts the retention time so that the same peptide is appropriately matched among all the runs. In peak apex alignment, the tops of the coeluting peaks have exactly the same retention time. Peak apex alignment is for cosmetic purposes only.
Abundance is the area under the curve for each trace. In the following figure, the Consensus Features page of the table associated with the selected peptide group shows the abundances displayed in the Chromatogram Traces view.
The Chromatogram Traces view can display three types of chromatograms: TIC, base peak, and Minora LC/MS peak trace. The processing workflow must include the Minora Feature Detector node to generate the latter two.
The Spectrum Selector node in the processing workflow creates the Specialized Traces page in the result file. It adds rows to the page for each input file for a base peak and TIC chromatogram. For more information on the Specialized Traces page, see The Specialized Traces page.
When the consensus workflow includes the Feature Mapper node, the Chromatogram Traces view also displays the chromatograms corrected for retention time. You can use this view to see how well the retention-time correction worked.
You can display the Chromatogram Traces view for items on the following pages of the result file. The information shown in the Chromatogram Traces view depends on the page that you select.
Page | Description |
|---|---|
MS/MS Spectrum Info page | Displays the overlaid traces of the LC/MS feature that are associated with the spectrum. An item on the MS/MS Spectrum Info page is usually associated with multiple PSMs and is therefore associated with multiple features. A bar indicates the time that the MS2 spectrum was acquired. |
LCMS Features page | Displays the trace of the selected LC/MS features. An LC/MS feature represents the height and area of the most abundant isotope of the quantified peptide. |
Consensus Features page | Displays the overlaid chromatograms for a specific feature across all the files. |
PSMs page | Displays overlaid traces of the LC/MS features associated with the selected PSMs. A PSM is associated with one feature. A bar indicates the time that the MS2 spectrum was acquired. |
Peptide Groups page | Displays the overlaid traces of the LC/MS features associated with the PSMs contained in the peptide group. Multiple charge states (for example, 2+, 3+) can result in more than one consensus feature. |
Specialized Traces page | Displays total ion chromatograms and base peak. |