You can use a Venn diagram to compare distinct and shared identifications for up to three groups of data. The available groups depend on the processing nodes that you used to generate the data in the result file. Typically, you might want to compare the distinct and shared protein or peptide identifications between the following:
- Different search engine nodes, for example, Sequest HT (2) and Sequest HT (8)
- Different search engine types, for example, Sequest HT and Mascot
- Different input files, for example, replicate A and replicate B
Procedure
- Select View > Distribution Charts, or select Distribution Charts
.
- Select the Venn Diagrams tab in the Report Item Distribution dialog box.
- From the Data Source list, select the type of data to display.
- The available result categories depend on the search engine nodes used in the workflow that generated the Report Item Distribution chart.
- From the 1. Group list, select the first data set to compare.
- From the menu in the Color A box, select the color to display this data set in.
- From the 2. Group list, select the second data set to compare.
- From the menu in the Color B box, select the color to display this data set in.
- (Optional) From the 3. Group list, select the third data set to compare.
- From the menu in the Color C box, select the color to display this data set in.
- Select Refresh.
- (Optional) Hide the Options pane by selecting Auto Hide
.
- The vertical icon shows the pane; the horizontal icon hides the pane. Point to Options to access the hidden Options pane.
- Use the options in the Options pane to adjust the colors, labels, and legends of the display. For information on these options, refer to Parameters on the Venn Diagrams view in the Proteome Discoverer Help.
- –or–
- In the Options pane of the Report Item Distribution chart, select Load to load the settings that you most recently saved.
- (Optional) To save all the settings that you have chosen, select Save in the Options pane.
- The new settings overwrite any previously saved settings. The application loads these new settings when you select Load.
Task | Procedure |
|---|---|
Return the settings to the original default settings | In the Options pane of the Report Item Distribution chart, select Factory Defaults. |
Copy a Venn diagram image | Procedure
|
Copy the points in a Venn diagram as text | Procedure
|
Save a Venn diagram as an image file | Procedure
|
Save the data from the Venn diagram in a text file | Procedure
|
Close the Report Item Distribution chart | Procedure
|