You can use pie chart to indicate the relative size of quantities of data.
Procedure
- In an open result file, select View > Distribution Charts, or select Distribution Charts
.
- Select the Pie Charts tab in the Report Item Distribution dialog box.
- From the Data Source list, select the type of data to display as a pie chart.
- Select Refresh to draw the chart.
- (Optional) Hide the Options pane by selecting Auto Hide
.
- The vertical icon shows the pane; the horizontal icon hides the pane. Point to Options to access the hidden pane.
- Use the options in the Options pane to change the type of chart from pie to doughnut and adjust the colors, labels, and legends of the display. For more information, see Parameters in the Pie Charts view in the Proteome Discoverer Help. The chart does not display any items that you have filtered out.
- –or–
- Select Load to load the settings that you most recently saved.
- (Optional) To save all the settings that you have chosen, select Save in the Options pane.
- The new settings overwrite any previously saved settings. The application loads these new settings when you select Load.
- Do any of the tasks listed in the following table.
Task | Procedure |
|---|---|
Return the settings to the original default settings | In the Options pane of the Report Item Distribution chart, select Factory Defaults. |
Copy a pie chart image | Procedure
|
Copy the points in a pie chart as text | Procedure
|
Save a pie chart as an image file | Procedure
|
Save the data from the pie chart in a text file | Procedure
|
Close the Report Item Distribution chart | Procedure
|