Bar charts plot categorical and ordinal data types in column charts that show the count of the data types as the column height. Although pie charts show the same data types as bar charts, percentage values in bar charts do not need to add up to 100 percent. This type of data display is especially useful for gene ontology annotations, because a protein is often assigned to more than one GO slim term; therefore, the fractions of the proteins assigned to the single GO slim terms do not add up to 100 percent.
The numerical data categories of the open results report that are available for the bar chart are listed in the Data Source selection box at the top of the Report Item Distribution view. The Options pane lists the different options that you can use to customize the bar chart. For information on these options, refer to the Parameters in the Bar Charts view Help topic. Pointing to the columns activates a tooltip with information about the data category.
Procedure
- In an open result file, select View > Distribution Charts or select Distribution Charts
.
- Select the Bar Charts tab in the Report Item Distribution view.
- From the Data Source list, select the type of data to display.
- Select Refresh to draw the chart.
- (Optional) Hide the Options pane by selecting Auto Hide
.
- The vertical icon shows the pane; the horizontal icon hides the pane. Point to Options to access the hidden pane.
- To adjust the colors, labels, and legends of the display, use the options in the Options pane. For information on these options, refer to Parameters in the Bar Charts view in the Proteome Discoverer Help.
- –or–
- In the Options pane of the Report Item Distribution chart, select Load to load the option settings that you most recently saved.
- The following figure is a bar chart that shows the Molecular Function counts. The tooltip shows the details of the data group.
- (Optional) To save your option settings, select Save in the Options pane.
- The new settings overwrite any previously saved settings. The application loads these new settings when you select Load.
- Do any of the tasks listed in the following table.
Task | Procedure |
|---|---|
Return the settings to the original default settings |
|
Copy a bar chart image | Procedure
|
Copy the points in a bar chart as text | Procedure
|
Save a bar chart as an image file | Procedure
|
Save the data from the bar chart in a text file | Procedure
|
Close the Report Item Distribution chart | Procedure
|