The following table lists the parameters in the Scatter Plots view.
Command or option | Description |
|---|---|
Options pane | |
Loads the Option pane settings that you saved with the Save command. | |
Saves the settings that you selected in the Options pane.
| |
Resets the settings of the options in the Options pane to the defaults that were in effect when you installed the application. | |
Chart Options | |
Horizontal Grid Lines | Determines whether the scatter plot displays horizontal grid lines and specifies the style of these lines.
|
Vertical Grid Lines | Determines whether the scatter plot displays vertical grid lines and specifies the style of these lines.
|
Axis Options | |
X-Axis Title | Specifies the label for the x axis. By default, this option uses the category selected from the X Data list as the x axis title. |
X Axis Type | Specifies the axis type (scale) of the x axis.
|
Y-Axis Title | Specifies the label for the y axis. By default, this option uses the category selected from the Y Data list as the y-axis title. |
Y Axis Type | Specifies the axis type (scale) of the y axis.
|
Z Axis Type | Specifies the axis type (scale) of the z axis.
|
Axis Title Font | Opens the Font dialog box so that you can specify the font used to denote the axis labels. For information, see the Font dialog box in the Proteome Discoverer Help. Default: 24-point Arial Select the arrow next to the Axis Label Font option to display aspects of the label font that you can change, which are as follows:
|
Axis Scale Font | Opens the Font dialog box so that you can specify the font used to denote the scale of the x and y axes. For information, see the Font dialog box in the Proteome Discoverer Help. Default: 18-point Arial. Select the arrow next to the Axis Scale Font option to display aspects of the label font that you can change, which are as follows:
|
Series Options | |
Points | Specifies the appearance of the items in the scatter plot that pass the used display filters. To display aspects of the points that you can change, select the arrow next to the Points option. The possible values are as follows:
|
Checked Points | Specifies the appearance of the items in the scatter plot that you selected in the results report. To display aspects of the selected points that you can change, select the arrow next to the Selected Points option. The available values are as follows:
|
Filtered-Out Points | Specifies the appearance of the items in the scatter plot filtered out by the current display filter. To display aspects of the selected points that you can change, select the arrow next to the Selected Points option. The available values are as follows:
|
Excluded Points | Specifies the appearance items in the scatter plot that were excluded in the consensus workflow. To display aspects of the excluded points that you can change, select the arrow next to the Excluded Points option. The available values are as follows:
|
Data Source | Displays the result category to plot the data from. The categories are as follows: Protein Groups, All Proteins, Peptide Groups, PSMs, or MS/MS Spectrum Info. |
X Data | Specifies the x-axis data set. |
Y Data | Specifies the y-axis data set. You can select the same types of data as for the x-axis data set. |
Z Data | Specifies the z-axis data set. You can select the same types of data as for the x-axis data set. |