The following table lists the parameters on the pages of the Peptide Spectrum Match Identification Details view.
Command or option | Description |
|---|---|
Options pane | |
Loads the last saved default fragment match settings for the selected mass analyzer/activation type combination. | |
Saves the current fragment match settings as the default fragment match settings for the selected mass analyzer/activation type combination. | |
Loads the original factory fragment match settings for all mass analyzer/activation type combinations. | |
Display Options area | |
Charge Detail Level | Selects the appropriate charge state to display. The values are as follows:
|
Annotation Threshold | Specifies a peak intensity limit. The application does not annotate peaks below this limit. This limit prevents the addition of too many labels to a spectrum, especially if the low-intensity peaks are possibly noise peaks.
|
Show Legend | Determines whether a legend consisting of colored lines that denote the displayed ion series appears at the bottom of each spectrum image. |
Show Reference Spectrum | Determines whether to display the reference spectrum from the reference spectrum library. |
Ref. Spectrum Source | Selects the reference spectrum source. The prediction is local or over the cloud.
|
Use Search Settings | Annotates the spectra using the match tolerances and ion series that you chose for the search node. |
Match Tolerances area | |
Mass Analyzer | Specifies which mass analyzer is used to obtain the spectrum. |
Mass Tolerance | Specifies the mass tolerance. The application annotates the spectrum with ions that it assigns within this mass tolerance. You can use daltons or millimass units. |
Fragments area | |
Activation Type | Specifies what type of activation produced the fragments. |
Ion Series area | Selects which series of fragment ions the application displays in the ion table and in the extracted data plot. By default, the spectrum displays the most relevant ion types for each activation type. See Displaying fragment ions for a list of the ions series that you can display. By default, the ion series are displayed in the following colors:
|
Neutral Losses area | Determines whether to display water or ammonia neutral losses in the ion table and in the extracted data plot. The application adds a column for each type of neutral loss ion that you select on the Neutral Losses page. For example, if you select the b and y ions in the Ion Series area and then choose the –NH3 checkbox, the application adds both a b–NH3 column and a y–NH3 column to the table on the Neutral Losses page. It also adds these ions to the extracted data plot. The application only calculates water neutral loss fragments if side chains contain the potential neutral loss groups of aspartic acid, glutamic acid, serine, or threonine. It only calculates ammonia neutral loss fragments if side chains contain the potential neutral loss groups of lysine, asparagine, glutamine, or arginine. |
Other area | Annotates the spectra with immonium ions or precursor ions. |
Peptide Summary pane | |
Description of spectra in a scroll box | Displays the following spectra information:
Below this information appears the following:
|
Fragment Matches pane | |
Value Type list | Specifies which unit is used to display the fragment ion masses in the ion table:
|
Ion table |
|
Fragment Spectrum pane | |
Extracted data plot | Shows the annotated spectrum with all the fragments that matched an expected fragment from in-silico digestion.
|