The Proteins page in a result file includes a Sequence Coverage column, which is automatically generated by the Peptide in Protein Annotation node. This column highlights in blue the parts of the protein sequence that the application identified. The column tooltip shows the number of amino acids composing the protein.
NOTE
You cannot export this column to a text file.
Sequence Coverage column on the Proteins page of a result file