The Modification List page within the Coverage page of the Protein Identification Details view lists all the identified PTMs that appear in the sequence table. For information on the columns on this page, see Parameters on the Coverage page of the Protein Identification Details view. You can adjust the length of the sequence motif before and after the modification site by using the Number of Amino Acids Displayed Before and After the Modification option.
Modification List page of the Coverage page within the Protein Identification Details view