When you use the Protein Annotation node in a consensus workflow to generate a result file, you can display maps of the chemical pathways identified for a protein by the Reactome pathway database and the Wiki Pathways database.
Procedure
- Open the result file.
- Select the Proteins tab.
- Select the protein of interest on the Proteins page.
- Select View > Pathway Map, or select Show the Biochemical Pathway Map
in the toolbar.
- The Pathway Map view opens. On the left is a list of all pathways assigned to the protein, and on the right is a pane showing the selected pathway.
Pathway Map view showing the selected biochemical pathway from the Wiki PATHWAY database - (Optional) Select a pathway identification in the left pane.
- The right pane displays the selected pathway.