In order to generate data distribution maps in your result file, you must create a workflow using the Data Distribution node (see Create a workflow).
Procedure
- To create a distribution map, ensure that the Data Distributions node is in the Post-Processing area of the Workflow Editor.
- Set the parameters of the Data Distributions node to display the appropriate columns in the output result file.
- For information about these parameters, see Data Distributions node in the Proteome Discoverer Help.