You can match likely proteins to your analyzed data and access isotope, chromatogram, and other graphical views. The views provide detailed graphical information about the selected PSMs and let you to perform a comparative analysis of your selected peptide or proteins. To move each view, see Manipulate search results views and Workflow Editor panes in the Proteome Discoverer Help.
Commands in the View menu provide access to the views on each page, or you can use the toolbar command buttons.
NOTE
The Quan Ratio Distributions chart (opened by the Quan Ratio Distributions command), Quan Channel Values chart (opened by the Quan Channel Values command), and Quantification Spectrum chart (opened by the Quantification Spectrum command) are described in Interpreting quantitative results. For access to these commands, you must open a results report containing quantification results.
Use the shortcut menu on each page of the result file to alter the details of that page in the result or to copy information.