You can specify biological replicates in a study. This information is required for calculating p-values with the Precursor Ions Quantifier node or the Reporter Ions Quantifier node.
Experiments can be divided into two groups, non-nested and nested.