The following table lists the parameters in the Sample Abundances view.
Parameter | Description |
|---|---|
Color By | Creates sample groups according to the study factors for quantification:
|
Filter By | Filters the data according to the following variables:
|
Data Source | Specifies the result category to plot the data from:
|
Transform Data | Determines whether the data appears in a linear scale or the log10 scale. Selecting this checkbox transforms the area counts to the log10 scale. |
Use Normalized Areas | Displays normalized chromatographic peak areas. The checkbox is available if the Reporter Ions Quantifier node or the Precursor Ions Quantifier node normalized the quantification data, but did not scale it. |
Graph | |
Groups (x axis) | Displays the name of the sample groups. |
LogArea/Area (109) (y axis) | Displays the area in a linear scale or in a log10 scale, depending on the setting of the Log-Transform Data parameter.
|
Rectangles | The rectangles represent the second and third quartiles for the data set. When you choose the Show Position tooltips command, placing the cursor over the rectangle displays the following information:
|
Circles | Represent outlier data points. When you choose the Show Position tooltips command, pointing to a data point displays the following information:
|
Legend | Displays the group colors. By default, the legend is hidden. Choose Show Legend from the shortcut menu to display it. |