You can generate a chart showing the spectrum used for quantification. This chart is available for every peptide with an associated quantification result. You must conduct a search with a workflow that includes a quantification node for this page to appear in the results report.
Procedure
- Select the appropriate tab in the result file.
- Select the row that interests you.
- Choose View > Quantification Spectrum, or select Quantification Spectrum
.