Procedure
- Select the Proteins or Peptide Groups tab in the result file.
- Select the row of the protein or peptide group that interests you.
- Choose View > Quan Channel Values, or select Quan Channel Values
.
- The following figure shows the grouped and ungrouped abundances on the Proteins page of the result file for label-free quantification.
Quantification Channel values chart showing grouped and ungrouped abundances for label-free quantification
The x axis of the chart shows the quantification channels, and the y axis shows the scaled abundance or summed S/N values, depending on whether you chose scaling in the Precursor Ions Quantifier node.
Task | Procedure |
|---|---|
Display the legend | Right-click the Quan Channel Values pane and choose Show Legend. |
Display grouped abundances | Right-click and choose Show Grouped Abundances. |
Display ungrouped abundances | Right-click and choose Show Ungrouped Abundances. |