The Quantification Channel Values chart for precursor ion quantification displays the area of the isotopes detected for the available quantification channels when you select a PSM or a quantification spectrum. When you select a protein or a peptide group, the chart displays the abundance of that item.