The Quantification Channel Values chart for reporter ion quantification displays the absolute intensity of the reporter ions detected for the available quantification channels when you select a PSM or a quantification spectrum. When you select a protein or a peptide group, the chart displays the abundance of the item.