The application counts the number of peptides that are only contained in a protein group and displays the value in the # Unique Peptides column on the Proteins page. The Proteome Discoverer application counts only the peptides that display a status of Selected or Unambiguous in the PSM Ambiguity column, because assessing the uniqueness of peptides that were not used to form protein groups has no relevance.